missing translation for 'onlineSavingsMsg'
Learn More

Pyrazinamide, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111016013
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016013 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016013 Supplier Merck Emd Millipore Supplier No. PHR1576500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 98-96-4
Linear Formula C5H5N3O
MDL Number MFCD00006132
Melting Point 189°C to 191°C
Synonym Pyrazinecarboxamide; Pyrazinamide; Pyrazinoic acid amide
Quantity 500 mg
Formula Weight 123.11
Grade Certified Reference Material