missing translation for 'onlineSavingsMsg'
Learn More

Pyrazinamide, 99%

Catalog No. AC157641000
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157641000 100 g Plastic bottle
AC157640100 10 g Plastic bottle
AC157640250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC157641000 Supplier Thermo Scientific Chemicals Supplier No. 157641000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 98-96-4 | C5H5N3O | 123.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-96-4
Molecular Formula C5H5N3O
Molecular Weight (g/mol) 123.11
MDL Number MFCD00006132
InChI Key IPEHBUMCGVEMRF-UHFFFAOYSA-N
Synonym pyrazinamide, pyrazinecarboxamide, zinamide, pyrazinoic acid amide, 2-pyrazinecarboxamide, aldinamide, aldinamid, pirazinamid, pyrazineamide, pyrafat
PubChem CID 1046
ChEBI CHEBI:45285
IUPAC Name pyrazine-2-carboxamide
SMILES C1=CN=C(C=N1)C(=O)N

Specifications

Melting Point 188°C to 191°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Quantity 100 g
Merck Index 15, 8067
Solubility Information Solubility in water: 15mg/mL. Other solubilities: 13.8mg/mL methanol- 5.7mg/mL abs. ethanol,3.8mg/mL isopropanol- 1.0mg/mL ether,0.01mg/mL isooctane- 7.4mg/mL chloroform
Formula Weight 123.11
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material Pyrazinamide, 99%
Show More Show Less
EINECSNumber 202-717-6
RTECSNumber UQ2275000
TSCA TSCA

RUO – Research Use Only