missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GVS PVDF-Plus™ Transfer Membrane
Description
- Superior strength: Can withstand aggressive handling or be used with automated equipment without breaking or tearing
- Low extractables: Ensures tests will be clean with consistent results
- Exceptional sensitivity: Detects low-level components
- Hydrophobic:For high protein binding
- Lot-to-lot consistency: Quality checks ensure consistent binding for dependable results every time
- BSA protein binding capacity is 125mg/cm2
- High range of chemical: Resistant to most commonly used chemicals compatible with chemically aggressive solvents
Specifications
Specifications
| Length (Metric) | 3 m |
| Width (Metric) | 15 cm |
| For Use With (Application) | Western blots, Immunoblotting, Solid phase assays, Amino acid or protein analyses |
| Binding Capacity | 125 mg/cm2 |
| Color | White |
| Format | Roll |
| Material | PVDF |
| Pore Size | 0.45 μm |