missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Purple Screw Caps
Description
- Purple Caps fit empty FastPrep™ tubes and are used in FastPrep™ systems.
- Add sample to empty tube, add Lysing Matrix E material and lysing buffer, and apply cap. Process using FastPrep™ instrument or other bead-beater to lyse, grind or homogenize sample.
- Purple caps are also found in the Fast DNA™ Spin Kit for Soil and Fast RNA™ Pro Soil Kit.
Spécifications
Spécifications
| Color | Purple |
| Volume (Metric) | 2 mL |
| Storage Requirements | Store at Room Temperature (15°C to 30°C) |
| Quantity | 1000 Pk. |
| Product Type | Screw Cap |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.