missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Purmorphamine

Numéro de catalogue. 455110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
455110 10 mg
455150 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 455110 Fournisseur Tocris Bioscience™ Code fournisseur 4551/10
Il en reste null

Smo receptor agonist

Purmorphamine is a smoothened (Smo) receptor agonist (EC50 ∼ 1 μM) and activates the Hedgehog pathway. Purmorphamine induces osteogenesis in mouse mesenchymal progenitor cells (C3H10T1/2). When combined with BMP-4, Purmorphamine can transdifferentiate pre-adipocytes (3T3-L1) and myoblasts (C2C12) into osteoblasts. Purmorphamine induces differentiation of multipotent mesenchymal progenitor cells into osteoblasts, and of spinal motor neurons from pluripotent human stem cells, and protects dopaminergic neurons in a mouse model of Parkinson's disease.

Identifiants chimiques

CAS 483367-10-8
Molecular Formula C31H32N6O2

Spécifications

Quantity 10 mg
Formula Weight 520.62
Molecular Weight (g/mol) 520.637
Chemical Name or Material Purmorphamine
Recommended Storage Store at -20°C