missing translation for 'onlineSavingsMsg'
Learn More

Purified Water, USP, Fisher Chemical™

Catalog No. W1220
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Plastic Bottle
PolyPac™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
W1220 20 L PolyPac™
W124 4 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. W1220 Supplier Thermo Fisher Scientific Supplier No. W1220
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
CAS Min % 100%
Boiling Point 100°C
CAS Max % 100%
Description Filtered to 0.2 μm
Grade USP
Packaging PolyPac™
Quantity 20 L
Physical Form Liquid
Chemical Name or Material Purified Water
Show More Show Less
Precautionary Statement
Note: Products are tested to meet the analytical requirements of the noted grade (USP, Ph. EUR or otherwise). Product test results are available on the Certificate of Analysis or upon request for the specific product.
 
Products are for research use or further manufacturing. Products are not for direct administration to humans or animals. It is the responsibility of the final formulator and end-user to qualify, test and/or validate each product for its intended use as an ingredient in the end product and in accordance with applicable regulations (including without limitation WHO-recommended Hand Rub Formulations (Hand Sanitizers) and/or FDA Policy for Preparation of Certain Alcohol-based Hand Sanitizer Products, as updated). Thermo Fisher Scientific expressly disclaims all non-specified warranties, expressed or implied, including the implied warranties of merchantability and fitness for a particular use.
ShelfLife 5 Years
Recommended Storage Room Temperature