missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PureProteome™ Protein G Magnetic Bead System

Catalog No. LSKMAGG10
Change view
Click to view available options
Quantity:
10 mL
2 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LSKMAGG10 10 mL
LSKMAGG02 2 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LSKMAGG10 Supplier MilliporeSigma™ Supplier No. LSKMAGG10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Requires

Magna Grip™ Rack, eight-well (Mfr. No. 20-400)

Eliminate variability and maximize recovery. Unlike conventional methods, which require centrifugation and careful aspiration, these beads are isolated using a magnetic rack. This allows for the total removal of buffers for complete recovery of beads and no sample dilution.

  • Bind 1.5 to 3.5μg of Rabbit IgG per μL of suspension
  • More than 6x the binding capacity of competitive magnetic beads
  • Total removal of buffers with no sample loss for consistent results
  • Beads are immobilized in seconds
  • Economical
  • Particle size: 10μm
  • Not recommended for Chromatin Immunoprecipitation (ChIP)

Serum depletion and immunoprecipitation assays

Specifications

Binding Property 2.5 to 3.5μg/μl
Content And Storage 2°C to 8°C
Particle Size 10 μm
Quantity 10 mL
For Use With (Application) Serum depletion, immunoprecipitation
Type Protein G Magnetic Bead System

Unless otherwise stated in the catalog or other company documentation accompanying the product(s), these products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.