missing translation for 'onlineSavingsMsg'
Learn More

Purell™ Hand Sanitizing Wipes Stainless Steel Stand Dispenser

Catalog No. 19043938
Change view
Click to view available options
Mounting:
Freestanding
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
19043938 Freestanding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19043938 Supplier GOJO Canada Inc Supplier No. 9115DS1C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High capacity floor stand dispenser for Purell™ wipes.

  • Weighted base keeps stand in place
  • Sleek and attractive Stainless Steel finish
  • Pre-installed SIMPUL™ nozzle technology, virtually eliminates roping, providing one wipe every time

Specifications

Product Type Dispenser Stand
Color Silver
Length (Metric) 10.25 cm
Width (English) 10.25 in.
Width (Metric) 10.25 cm
Height (English) 28 in.
Height (Metric) 28 cm
Holds Hand Sanitizer
Material Plastic, Metal
Mounting Freestanding
No. of Compartments 1
Product Style Top Loading
For Use With (Equipment) PURELL™ Hand Sanitizing Wipes
Quantity 1 Ea.
Show More Show Less