Learn More
Gente™ PureFlo 3000 Respirator with Open Frame and Clear Visor
Numéro de catalogue. 17720125
Change view
Click to view available options
:
Gente™ Respirator with Open Frame
Numéro de catalogue.
17720125 Gente™ Respirator with Open Frame
1 options
Numéro de catalogue. 17720125 Fournisseur Gente™ Code fournisseur PF3000F2SB03NBC
Il en reste null
PureFlo 3000 respirator with open frame and clear visor. Standard face seal.
  • PureFlo 3000 respirator with open frame and clear visor.
  • Standard face seal with no battery charger.
Spécifications
Assigned Protection Factor (APF) 25
Cartridge/Filter PARTICULATE
Color Gray
Product Type PAPR Assembly, No Headtop
Size One Size
Filter Efficiency (NIOSH) HEPA
Intrinsic Safety (IS) Rating No
IP Rating IP54