Learn More
Gente™ PureFlo 3000 Respirator with Open Frame
Description
- PureFlo 3000 respirator with open flame and Tychem™ quality coated hood.
- No battery charger included.
Spécifications
Spécifications
| Assigned Protection Factor (APF) | 25 |
| Cartridge/Filter | PARTICULATE |
| Color | Gray |
| Product Type | PAPR Assembly, No Headtop |
| Size | One Size |
| Intrinsic Safety (IS) Rating | No |
| IP Rating | IP54 |