Learn More
Mopec Pure Path™ Room Filtration Module
Catalog No. 22444007
Change view
Click to view available options
Includes:
2 replaceable 40lb (18kg) potassium permanganate and carbon mixed filters
Catalog No. Includes
22444007 2 replaceable 40lb (18kg) potassium permanganate and carbon mixed filters
1 options
Catalog No. 22444007 Supplier Mopec Supplier No. BF840
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Possesses the horsepower to rein in formalin overexposure and maintain a safe laboratory environment. Mopec™ Pure Path™ Room Filtration Module is a heavy duty auxiliary air filtration unit designed to neutralize residual formalin fumes in the busiest of laboratories.
  • Constructed of stainless steel
  • Equipped with a 9 foot line cord with hospital grade plug
  • Casters enable easy positioning throughout any lab, allowing easy filtration of problem areas
Warranty
1 Year Warranty
Specifications
Product Type Filtration Module
Includes 2 replaceable 40lb (18kg) potassium permanganate and carbon mixed filters