Learn More
Fisherbrand™ Punched Aluminum Test Tube Rack
Catalog No. 01189160
Change view
Click to view available options
Size:
24 Slot
40 Slot
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
01189160 40 Slot
01189161 24 Slot
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01189160 Supplier Fisherbrand™ Supplier No. 10401277
Only null left
This Punched Aluminum Test Tube Rack features a Z-shape for holding test tubes. Three configurations available: 24, 25, and 40 slots.
  • Aluminum construction
  • Anti-corrosive
  • Low profile and well balanced
  • Available in three configurations: 24, 25, and 40 slots
Specifications
Certifications/Compliance ISO, GMP
Autoclavable Autoclavable
Color Silver
Holds Test Tubes
Material Stainless Steel
Shape Z-shaped
Stackable Yes, When Empty
Sterility Non-sterile
Size 40 Slot
Product Type Test Tube Rack