missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTGES2 Monoclonal Antibody (K06332_11H12), Invitrogen™

Numéro de catalogue. MA556217
Modifier l'affichage
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA556217 200 μL
MA555837 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA556217 Fournisseur Invitrogen™ Code fournisseur MA556217
Il en reste null

Mouse Monoclonal Antibody

Sequence of this protein is as follows: AAQLSLSSRL QLTLYQYKTC PFCSKVRAFL DFHALPYQVV EVNPVRRAEI KFSSYRKVPI LVAQEGESSQ QLNDSSVIIS ALKTYLVSGQ PLEEIITYYP AMKAVNEQGK EVTEFGNKYW LMLNEKEAQQ VYGGKEARTE EMKWRQWADD WLVHLISPNV YRTPTEALAS FDYIVREGKF GAVEGAVAKY MGAAAMYLIS KRLKSRHRLQ DNVREDLYEA ADKWVAAVGK DRPFMGGQKP NLADLAVYGV LRVMEGLDAF DDLMQHTHIQ PWYLRVERAI

The PTGES2 gene encodes for the enzyme prostaglandin E synthase 2 (PTGES2), which is crucial for the biosynthesis of prostaglandin E2 (PGE2) from arachidonic acid. This enzyme operates downstream of the cyclooxygenase (COX) pathway, particularly interacting with COX-1 and COX-2 enzymes. PTGES2 is involved in various physiological and pathological processes by generating PGE2, which functions in inflammation, fever generation, and pain. Notably, PTGES2 is a membrane-associated protein and is distinct in its role compared to other prostaglandin synthases due to its specific functional and regulatory properties. Its activity is essential in fine-tuning the levels of PGE2 in tissues, impacting numerous signaling cascades, including those involved in immune responses and vascular functions.

Spécifications

Antigen PTGES2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K06332_11H12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Ptges2
Gene Accession No. Q9H7Z7
Gene Alias 0610038H10Rik; C79137; C9orf15; FLJ14038; gamma-interferon-activated transcriptional element-binding factor 1; GATE-binding factor 1; Gbf1; GBF-1; membrane-associated prostaglandin E synthase 2; membrane-associated prostaglandin E synthase-2; MGC11289; Microsomal prostaglandin E synthase 2; microsomal prostaglandin E synthase-2; mPGE synthase-2; Mpges2; mPGES-2; PGES2; Prostaglandin E synthase 2; Prostaglandin E synthase 2 truncated form; Prostaglandin-H(2) E-isomerase; PTGES2; RP11-395P17.2
Gene Symbols Ptges2
Host Species Mouse
Immunogen Recombinant human PTGES2 (immunogen range is 91-370).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80142
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Afficher plus de résultats Afficher moins de résultats