missing translation for 'onlineSavingsMsg'
Learn More

PTFE rod, 16mm (0.63 in.) dia, Thermo Scientific™

Catalog No. AA45173BV
Change view
Click to view available options
Quantity:
30 cm
Packaging:
Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA45173BV 30 cm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45173BV Supplier Thermo Scientific Chemicals Supplier No. 45173BV

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 9002-84-0
Molecular Formula (C2F4)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084455
SMILES FC(F)(-*)C(F)(F)-*

Specifications

Packaging Tube
Quantity 30 cm
Chemical Name or Material PTFE rod
TSCA U
Recommended Storage Ambient temperatures