Learn More
Sciencix Sciencix Deuterium Lamp, Intelligent Chip, 2000 hr Lifetime, 5 pcs
Numéro de catalogue. NC2055002
Encompass
The Sciencix Deuterium Lamp is engineered for high-performance liquid chromatography (HPLC) detectors, providing a reliable and stable light source with a lifetime of 2000 hr. Ideal for scientific applications, this lamp features an intelligent chip, ensuring compatibility with Waters ACQUITY systems while being a high-quality alternative to OEM parts. Its lifetime warranty against manufacturing defects adds further value, ensuring customers can trust its performance and durability in demanding environments.
Numéro de catalogue. NC2055002 Fournisseur Sciencix Code fournisseur CTS21065
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The Sciencix Deuterium Lamp is engineered for high-performance liquid chromatography (HPLC) detectors, providing a reliable and stable light source with a lifetime of 2000 hr. Ideal for scientific applications, this lamp features an intelligent chip, ensuring compatibility with Waters ACQUITY systems while being a high-quality alternative to OEM parts. Its lifetime warranty against manufacturing defects adds further value, ensuring customers can trust its performance and durability in demanding environments.