Learn More
Anton Paar Pt100 temperature sensor for ViscoQC 100 & ViscoQC 300
Numéro de catalogue. NC1818584
Encompass
Optional Pt100 sensor to monitor the sample temperature. The sensor includes a permanently connected cable for connection to ViscoQC. A guide and a release knob secure the plug against faulty connection and accidental loosening. Comes with a metal clip (mounting clamp) for fixing the Pt100 sensor on the rim of open sample vessels or on the L/R spindle guard.Suitable for use with Anton Paar rotational viscometers ViscoQC 100 L item NC1818559, ViscoQC 100 R item NC1818567, ViscoQC 100 H item NC1818569, ViscoQC 300 L item NC1818572, ViscoQC 300 R item NC1818576, ViscoQC 300 - H item NC1818578.
Numéro de catalogue. NC1818584 Fournisseur Anton Paar Code fournisseur ANP 191705
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Optional Pt100 sensor to monitor the sample temperature. The sensor includes a permanently connected cable for connection to ViscoQC. A guide and a release knob secure the plug against faulty connection and accidental loosening. Comes with a metal clip (mounting clamp) for fixing the Pt100 sensor on the rim of open sample vessels or on the L/R spindle guard.Suitable for use with Anton Paar rotational viscometers ViscoQC 100 L item NC1818559, ViscoQC 100 R item NC1818567, ViscoQC 100 H item NC1818569, ViscoQC 300 L item NC1818572, ViscoQC 300 R item NC1818576, ViscoQC 300 - H item NC1818578.