missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMD1 Monoclonal Antibody (K70027_1F5), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555734
Description
Sequence of this protein is as follows: LNKDLKMPKV QYKSNCKPST FAYPAPLEVP KEKEKEKVST AVLSITAKAK KKEKEKEKKE EEKMEVDEAE KKEEKEKKKE PEPNFQLLDN PARVMPAQLK VLTMPETCRY QPFKPLSIGG IIILKDTSED
Proteolytic degradation is critical to the maintenance of appropriate levels of short-lived and regulatory proteins as important and diverse as those involved in cellular metabolism, heat shock and stress response, antigen presentation, modulation of cell surface receptors and ion channels, cell cycle regulation, transcription, and signalling factors. The ubiquitin-proteasome pathway deconstructs most proteins in the eukaryotic cell cytosol and nucleus. Others are degraded via the vacuolar pathway which includes endosomes, lysosomes, and the endoplasmic reticulum. The 26S proteasome is an ATP-dependent, multisubunit (~31), barrel-shaped molecular machine with an apparent molecular weight of ~2.5 MDa. It consists of a 20S proteolytic core complex which is crowned at one or both ends by 19S regulatory subunit complexes. The 19S regulatory subunits recognize ubiquitinated proteins and play an essential role in unfolding and translocating targets into the lumen of the 20S subunit. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis, Angelman's syndrome, and Liddle syndrome.Identifiants chimiques
| PSMD1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 2410026J11Rik; 26S proteasome non-ATPase regulatory subunit 1; 26S proteasome p112 subunit; 26S proteasome regulatory subunit RPN2; 26S proteasome regulatory subunit S1; 26S proteasome subunit p112; 26S proteasome, subunit p112; MGC133040; MGC133041; P112; proteasome (prosome, macropain) 26S subunit, non-ATPase, 1; proteasome 26S non-ATPase subunit 1; proteasome 26S subunit, non-ATPase 1; Psmd1; Rpn2; S1 | |
| Recombinant human PSMD1 (immunogen range is 791-920). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PSMD1 |
| K70027_1F5 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q99460 | |
| 5707 | |
| IgG2b | |
| Protein A | |
| Antibody |
Spécifications
| PSMD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q99460 | |
| PSMD1 | |
| Recombinant human PSMD1 (immunogen range is 791-920). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| K70027_1F5 | |
| Unconjugated | |
| PSMD1 | |
| 2410026J11Rik; 26S proteasome non-ATPase regulatory subunit 1; 26S proteasome p112 subunit; 26S proteasome regulatory subunit RPN2; 26S proteasome regulatory subunit S1; 26S proteasome subunit p112; 26S proteasome, subunit p112; MGC133040; MGC133041; P112; proteasome (prosome, macropain) 26S subunit, non-ATPase, 1; proteasome 26S non-ATPase subunit 1; proteasome 26S subunit, non-ATPase 1; Psmd1; Rpn2; S1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 5707 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |