missing translation for 'onlineSavingsMsg'
Learn More

Pseudopelletierine, Thermo Scientific Chemicals

Numéro de catalogue. AC463980010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC463980010 1 g
AC463980050 5 g
AC463980250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC463980010 Fournisseur Thermo Scientific Chemicals Code fournisseur 463980010
Il en reste null

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 552-70-5
Molecular Formula C9H15NO
Molecular Weight (g/mol) 153.23
InChI Key RHWSKVCZXBAWLZ-UHFFFAOYNA-N
IUPAC Name 9-methyl-9-azabicyclo[3.3.1]nonan-3-one
SMILES CN1C2CCCC1CC(=O)C2

Spécifications

Quantity 1 g
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 153.22
Chemical Name or Material Pseudopelletierine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

EINECSNumber 209-021-1

RUO – Research Use Only