missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ PRP-X100 Anion Exchange HPLC Columns: PEEK
Polymeric column packing for separation of inorganic and organic anions from 10 to 500 ppm
$1259.36 - $1681.77
Specifications
| For Use With | Analysis of Fluoride, Carbonate, Chloride, Nitrite, Bromide, Nitrate, Phosphate, Sulfate and Other Ions |
|---|---|
| Max. Pressure | 5000 psi (345 bar) |
| Mobile Phase Compatibility | pH 1 to 13, 0 to 100% aqueous, organic modifier |
| pH | 1 to 13 |
| Pore Size | 100 Å |
| Catalog Number | Mfr. No. | Particle Size | Flow Rate | Column Format | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Particle Size | Flow Rate | Column Format | Price | Quantity | |||
|
14-816-71
|
Hamilton™
HAM79354 |
10 μm | 8 mL/min | 150 x 4.6 mm |
Each for $1,259.36
|
|
|||
|
13-622-555
|
Hamilton™
HAM79174 |
5 μm | 1 mL/min | 150 x 4.6 mm L x ID |
Each for $1,442.92
|
|
|||
|
14-816-109
|
Hamilton™
HAM79455 |
10 μm | 1 mL/min | Analytical Column |
Each for $1,585.09
|
|
|||
|
13-622-556
|
Hamilton™
HAM79181 |
5 μm | 1 mL/min | 250 x 4.6 mm L x ID |
Each for $1,681.77
|
|
|||
Description
Easy Ion Determination
- PRP-X100 ion chromatography columns are the easy way to separate inorganic ions.These columns can be used with any existing HPLC or ion chromatograph to easily determine anions in almost any sample matrix including air, water and soil.
- Ion chromatography greatly reduces sample pretreatment and improves the accuracy and precision of results when using wet chemical or colorimetric methods
- Use to separate ions at concentrations from 1 ppm to 1,000 ppm without suppression or elaborate preconcentration techniques
- Highly inert polymeric support resists chemical attack from organic solvents and aqueous buffers (0-100% aqueous or organic; pH 1 to 13), effectively lengthening column life. If column performance should deteriorate (peak broadening and a loss of symmetry), the regeneration protocol will usually return chromatography to its original state.
- Easily separate difficult anions such as cyanide, borate and silicate at high pH (11.5). The polymeric packing is stable from pH 1 to 13, so a single column can be used for the analysis of both common and difficult anions.
- Conductivity detection works by measuring the increased conductance of the eluting sample ions relative to the background conductance
Specifications
| Analysis of Fluoride, Carbonate, Chloride, Nitrite, Bromide, Nitrate, Phosphate, Sulfate and Other Ions | |
| pH 1 to 13, 0 to 100% aqueous, organic modifier | |
| 100 Å | |
| PRP-X100 | |
| Ion Exchange | |
| PEEK |
| 5000 psi (345 bar) | |
| 1 to 13 | |
| 5°C to 60°C | |
| PSDVB/Trimethylammonium | |
| Ion Exchange | |
| Column |