missing translation for 'onlineSavingsMsg'
Learn More

Protocol™ Zinc Formalin, Fisherbrand™

Catalog No. 11002225
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002225 5 gal.
11002224 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002225 Supplier Statlab Supplier No. 11002225

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

Ready-to-use buffered zinc formalin in bulk packaging.

  • Especially recommended for use in immunohistochemistry, as zinc formalin preserves more immunoreactivity than standard NBF.
  • Used as a fixative solution that provides excellent preservation of tissue morphology, particularly nuclear and cytoplasmic components, by increasing the rigidity of cellular structures

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Formalin
Quantity 5 gal.
Solution Type Formalin
Form Solution

Made in the USA