missing translation for 'onlineSavingsMsg'
Learn More

Protocol™ SafeClear Xylene Substitute, Fisherbrand™

Catalog No. 11002220
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002220 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002220 Supplier Statlab Supplier No. 11002220

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

100% aliphatic hydrocarbon for processing and staining.

  • Xylene Substitute
  • Higher permissible exposure limit than xylene
  • Virtually no odor
  • Flash point 104°F
  • Exceptionally high purity

Specifications

Product Type Xylene Substitute
Quantity 1 gal.
Form Solution

Made in the USA