missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Prothionamide | |
| 143°C | |
| C9H12N2S | |
| 5 g | |
| VRDIULHPQTYCLN-UHFFFAOYSA-N | |
| 2-propylpyridine-4-carbothioamide | |
| 666418 | |
| ≥98.0% (HPLC,T) |
| 14222-60-7 | |
| Yellow | |
| MFCD00464119 | |
| 2-Propylpyridine-4-carbothioamide, 2-Propylthioisonicotinamide | |
| CCCC1=NC=CC(=C1)C(=S)N | |
| 180.269 | |
| 180.27 | |
| Crystalline Powder |
Chemical Identifiers
| 14222-60-7 | |
| 180.269 | |
| VRDIULHPQTYCLN-UHFFFAOYSA-N | |
| 666418 | |
| CCCC1=NC=CC(=C1)C(=S)N |
| C9H12N2S | |
| MFCD00464119 | |
| 2-Propylpyridine-4-carbothioamide, 2-Propylthioisonicotinamide | |
| 2-propylpyridine-4-carbothioamide |
Safety and Handling
RTECSNumber : NS0650000
TSCA : No