accessibility menu, dialog, popup

UserName

CAS RN 14222-60-7

CH3NNH2S

CAS RN 14222-60-7

IUPAC Name: 2-propylpyridine-4-carbothioamide
Synonyms: protionamide
Molecular Weight (g/mol): 180.27
Molecular Formula: C9H12N2S
InChi Key: VRDIULHPQTYCLN-UHFFFAOYSA-N
SMILES: CCCC1=CC(=CC=N1)C(N)=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results