missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prothionamide 98.0+%, TCI America™
$80.85 - $263.75
Chemical Identifiers
| CAS | 14222-60-7 |
|---|---|
| Molecular Formula | C9H12N2S |
| Molecular Weight (g/mol) | 180.269 |
| MDL Number | MFCD00464119 |
| InChI Key | VRDIULHPQTYCLN-UHFFFAOYSA-N |
| Synonym | 2-Propylpyridine-4-carbothioamide, 2-Propylthioisonicotinamide |
| PubChem CID | 666418 |
| IUPAC Name | 2-propylpyridine-4-carbothioamide |
| SMILES | CCCC1=NC=CC(=C1)C(=S)N |
Chemical Identifiers
| 14222-60-7 | |
| 180.269 | |
| VRDIULHPQTYCLN-UHFFFAOYSA-N | |
| 666418 | |
| CCCC1=NC=CC(=C1)C(=S)N |
| C9H12N2S | |
| MFCD00464119 | |
| 2-Propylpyridine-4-carbothioamide, 2-Propylthioisonicotinamide | |
| 2-propylpyridine-4-carbothioamide |
Specifications
| 14222-60-7 | |
| Yellow | |
| MFCD00464119 | |
| 2-Propylpyridine-4-carbothioamide, 2-Propylthioisonicotinamide | |
| CCCC1=NC=CC(=C1)C(=S)N | |
| 180.269 | |
| 180.27 | |
| Crystalline Powder |
| 143°C | |
| C9H12N2S | |
| 5 g | |
| VRDIULHPQTYCLN-UHFFFAOYSA-N | |
| 2-propylpyridine-4-carbothioamide | |
| 666418 | |
| ≥98.0% (HPLC,T) | |
| Prothionamide |
Safety and Handling
RTECSNumber : NS0650000
TSCA : No