missing translation for 'onlineSavingsMsg'
Learn More

ProteoMass™ LTQ/FT-Hybrid ESI Negative Mode Calibration Mix, MilliporeSigma™ Supelco™

Catalog No. 111016629
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016629 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016629 Supplier Merck Emd Millipore Supplier No. MSCAL61EA
The ProteoMass™ ESI Calibration Kit for LTQ based Hybrids, Negative Ion Mode Calibration Solution, enables calibration of Thermo Scientific™ hybrid instruments equipped with ESI source in negative ion mode. It covers a mass range of m/z 265 to m/z 1880, and is therefore usable for calibrations between m/z 50 and 2000.

Specifications

CAS 75-05-8
Product Line ProteoMass
Flash Point 5.6°C - closed cup
Shipping Condition Dry Ice
Concentration Varied Conc.
Type MALDI-MS Calibration Mix
Quantity 1 Ea.

ProteoMass is a trademark of Sigma-Aldrich Co. LLC