missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ProteoExtract ™Phosphopeptide Enrichment TiO2 Kit

Catalog No. 5397221KIT
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5397221KIT 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5397221KIT Supplier MilliporeSigma™ Supplier No. 5397221KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specialized extraction kit useful tool for enrichment of phosphorylated peptides from complex samples

  • Useful tool for enrichment of phosphorylated peptides from complex samples by a highly selective Titanium oxide solid phase
  • It enhances the ability to routinely identify and characterize large numbers of phosphorylated species within complex protein mixtures taking advantage of a novel titanium dioxide material
  • Captured and eluted phosphopeptide fractions are ready for MALDI- and ESI- mass spectrometry analysis

Specifications

Quantity 1 kit
Format
  • Phosphopeptide enrichment
  • Form: 100 Isolations
Kit Contents TiO2 phosphobind resin, 10X TiO2 phosphobind buffer, Wash buffer 1, Wash buffer 2, TiO2 Elution buffer, Dihydroxybenzoic acid, and user protocol
Content And Storage Store at+2°C to+8°C