Learn More
New England Biolab Ltd Proteinase K, Molecular Biology Grade 2 x 1 ml vials
Numéro de catalogue. NC0547027
Encompass
Proteinase K is a subtilisin-related serine protease that hydrolyzes a variety of peptide bonds and is frequently used to cleanup enzymatic reactions or cell lysates.
Numéro de catalogue. NC0547027 Fournisseur New England Biolab Ltd Code fournisseur P8107S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Proteinase K is a subtilisin-related serine protease that hydrolyzes a variety of peptide bonds and is frequently used to cleanup enzymatic reactions or cell lysates.