missing translation for 'onlineSavingsMsg'
Learn More

Protein Standard Set, MilliporeSigma™ Supelco™

Catalog No. 111016034
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016034 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016034 Supplier Merck Emd Millipore Supplier No. P54941SET
Only null left

Specifications

Solution Contains 0.05% sodium azide as preservative
Physical Form Neat
Type Analytical Standard
Quantity 1 Ea.
Grade Analytical Standard