missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Protein Kinase Inhibitor

Catalog No. AAJ61687MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61687MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61687MCR Supplier Thermo Scientific Chemicals Supplier No. J61687MCR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97161-97-2 | C27H21N3O5, C27H21N3O5 | 467.48 g/mol

Inhibitor of protein kinase A

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97161-97-2
Molecular Formula C27H21N3O5
Synonym K252a

Specifications

Quantity 1 mg
Description Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Inhibitor of protein kinase A
Formula Weight 467.47
Form Powder

RUO – Research Use Only