missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Protein A-HRP

Catalog No. 101023
Encompass
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
101023 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 101023 Supplier Invitrogen™ Supplier No. 101023

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protein A-HRP

Protein A from Staphylococcus aureus, Cowan I Strain conjugated with highly purified horseradish peroxidase (RZ > 3.0).

The product is liquid containing 10 mM phosphate, 0.15 M NaCl buffer, pH 7.4, 1% ovalbumin, 40% glycerol, and 0.1% proclin.

Specifications

Conjugate HRP
Quantity 2 mL
Form Solution
Protein Subtype Protein A
Content And Storage Store at 2-8°C
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.