missing translation for 'onlineSavingsMsg'
Learn More

Propyphenazone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014259
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014259 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014259 Supplier Merck Emd Millipore Supplier No. PHR13891G
Propyphenazone is a nonsteroidal drug exhibiting anti-inflammatory properties. It is widely used as a mild anesthetic medicine. It belongs to the pyrazolone class of compounds and has been posited to elicit an allergic or pseudo allergic reaction.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 479-92-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C14H18N2 O
MDL Number MFCD00053368
Synonym 4-Isopropylantipyrine; 1,2-Dihydro-1,5-dimethyl-4-(1-methylethyl)-2-phenyl-3 H-pyrazol-3-one; 1,2-Dihydro-4-isopropyl-1,5-dimethyl-2-phenyl-3 H-pyrazol-3-one; Isopropylphenazone; Propyphenazone
Type Certified Reference Material
Quantity 1 g
Formula Weight 230.31
Grade Certified Reference Material