missing translation for 'onlineSavingsMsg'
Learn More

Propyne (ca. 4% in N,N-Dimethylformamide, ca. 1.0mol/L), TCI America™

Catalog No. P2152100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2152100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2152100ML Supplier TCI America Supplier No. P2152100ML
Only null left

Chemical Identifiers

CAS 74-99-7
Molecular Formula C3H4
Molecular Weight (g/mol) 40.065
MDL Number MFCD00036235
InChI Key MWWATHDPGQKSAR-UHFFFAOYSA-N
Synonym Methylacetylene
PubChem CID 6335
ChEBI CHEBI:48086
IUPAC Name prop-1-yne
SMILES CC#C

Specifications

Color Yellow
Quantity 100 mL
UN Number 1992
Formula Weight 40.07
Physical Form Liquid
Chemical Name or Material Propyne (ca. 4% in N,N-Dimethylformamide, ca. 1.0mol/L)
RTECSNumber UK4250000
TSCA Yes
Recommended Storage Refrigerator