missing translation for 'onlineSavingsMsg'
Learn More

Propylene Oxide 99.0+%, TCI America™

Catalog No. E001625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E001625ML 25 mL
E0016500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E001625ML Supplier TCI America Supplier No. E001625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-56-9
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00005126
InChI Key GOOHAUXETOMSMM-UHFFFAOYNA-N
Synonym propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
PubChem CID 6378
ChEBI CHEBI:38685
IUPAC Name 2-methyloxirane
SMILES CC1CO1

Specifications

Color Colorless
Boiling Point 34°C
Quantity 25 mL
UN Number 1280
Formula Weight 58.08
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Propylene Oxide
EINECSNumber (2)-0219
RTECSNumber TZ2975000
TSCA Yes