missing translation for 'onlineSavingsMsg'
Learn More

(±)-Propylene Oxide, Analytical Standard, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020591
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020591 1 mL
111019167 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020591 Supplier Merck Emd Millipore Supplier No. 566711MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 75-56-9
Vapor Pressure 8.59 psi (20°C)
Vapor Density 2 (vs. air)
Density 0.83 g/mL (at 25°C)
Physical Form Neat
Linear Formula C3H6O
MDL Number MFCD00005126
Melting Point -112°C
Boiling Point 34°C
Synonym (+/-)-Methyloxirane; 1,2-Epoxypropane
Quantity 1 mL
Formula Weight 58.08
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less