missing translation for 'onlineSavingsMsg'
Learn More

Propylcyclohexane 98.0+%, TCI America™

Numéro de catalogue. P068125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P068125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. P068125ML Fournisseur TCI America Code fournisseur P068125ML
Il en reste null

Identifiants chimiques

CAS 1678-92-8
Molecular Formula C9H18
Molecular Weight (g/mol) 126.243
MDL Number MFCD00013770
InChI Key DEDZSLCZHWTGOR-UHFFFAOYSA-N
Synonym 1-Cyclohexylpropane
PubChem CID 15505
IUPAC Name propylcyclohexane
SMILES CCCC1CCCCC1

Spécifications

Melting Point -95°C
Color Colorless
Boiling Point 157°C
Quantity 25 mL
UN Number 3295
Formula Weight 126.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propylcyclohexane