missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Éther propylique 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -123°C |
| Color | Incolore |
| Boiling Point | 90°C |
| Quantity | 500 mL |
| UN Number | 2384 |
| Formula Weight | 102.18 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Propyl Ether |
Sécurité et manipulation
| EINECSNumber | (2)-0362 |
| RTECSNumber | UJ5125000 |
| TSCA | Yes |