missing translation for 'onlineSavingsMsg'
Learn More

Propyl Acetate 98.0+%, TCI America™

Catalog No. A004425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A004425ML 25 mL
A0044500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A004425ML Supplier TCI America Supplier No. A004425ML

Chemical Identifiers

CAS 109-60-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009372
InChI Key YKYONYBAUNKHLG-UHFFFAOYSA-N
Synonym n-propyl acetate, acetic acid, propyl ester, propyl ethanoate, 1-acetoxypropane, n-propyl ethanoate, 1-propyl acetate, acetic acid n-propyl ester, octan propylu, propylacetate, acetic acid propyl ester
PubChem CID 7997
ChEBI CHEBI:40116
IUPAC Name propyl acetate
SMILES CCCOC(C)=O

Specifications

Color Colorless
Boiling Point 101°C
Quantity 25 mL
UN Number 1276
Formula Weight 102.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Acetate
EINECSNumber (2)-0727
RTECSNumber AJ3675000
TSCA Yes