missing translation for 'onlineSavingsMsg'
Learn More

Propyl Acetate, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023052
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023052 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023052 Supplier Merck Emd Millipore Supplier No. RTC00009420ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 109-60-4
Melting Point -95°C (literature)
Density 0.888 g/mL (at 25°C (literature))
Boiling Point 102°C (literature)
Flash Point 14°C (Not Applicable)
Molecular Formula C5H10O2
Refractive Index n20/D 1.384 (literature)
Linear Formula CH3COOCH2CH2CH3
Quantity 20 mL
MDL Number MFCD00009372
Synonym Propyl acetate
Vapor Density 3.5 (vs. air)
Vapor Pressure 25 mmHg (20°C)
Formula Weight 102.13
Grade Certified Reference Material
Show More Show Less