missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Propranolol hydrochloride, 99%

Numéro de catalogue. p-7029206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAH2664506 5 g
AAH2664514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAH2664506 Fournisseur Thermo Scientific Chemicals Code fournisseur H2664506
Il en reste null

CAS: 318-98-9 | C16H22ClNO2 | 295.807 g/mol

A beta-adrenergic blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 318-98-9
Molecular Formula C16H22ClNO2
Molecular Weight (g/mol) 295.807
MDL Number MFCD00012558
InChI Key ZMRUPTIKESYGQW-UHFFFAOYSA-N
Synonym propranolol hydrochloride, propranolol hcl, inderal, avlocardyl, herzbase, inderalici, naprilin, pronovan, dociton, ikopal
PubChem CID 62882
ChEBI CHEBI:8500
IUPAC Name 1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride
SMILES CC(C)NCC(COC1=CC=CC2=CC=CC=C21)O.Cl

Spécifications

Melting Point 163°C to 165°C
Quantity 5 g
Merck Index 14,7840
Formula Weight 295.8
Percent Purity 99%
Chemical Name or Material (+/-)-Propranolol hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 206-268-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only