missing translation for 'onlineSavingsMsg'
Learn More

Propionyl bromide, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 392361000

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC392361000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

598-22-1
136.98
RIBFXMJCUYXJDZ-UHFFFAOYSA-N
69018
CCC(=O)Br
C3H5BrO
MFCD00000116
propionyl bromide, propanoyl-bromide, propionylbromide, propanyl bromide, acmc-209mg3, ksc270q9p, ribfxmjcuyxjdz-uhfffaoysa, inchi=1/c3h5bro/c1-2-3 4 5/h2h2,1h3
propanoyl bromide

Specifications

Propionyl bromide
1.5200g/mL
52°C
94% min. (GC)
C3H5BrO
CH3CH2COBr
MFCD00000116
propionyl bromide, propanoyl-bromide, propionylbromide, propanyl bromide, acmc-209mg3, ksc270q9p, ribfxmjcuyxjdz-uhfffaoysa, inchi=1/c3h5bro/c1-2-3 4 5/h2h2,1h3
RIBFXMJCUYXJDZ-UHFFFAOYSA-N
propanoyl bromide
69018
95%
598-22-1
104°C
Authentic
Glass bottle
1.4525 to 1.4575 (20°C, 589nm)
100g
1.52
Solubility in water: reacts violently
CCC(=O)Br
136.98
136.98
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-924-

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...