Learn More
Propionyl bromide, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 392360250
| Quantity | 25g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
| C3H5BrO | |
| MFCD00000116 | |
| propionyl bromide, propanoyl-bromide, propionylbromide, propanyl bromide, acmc-209mg3, ksc270q9p, ribfxmjcuyxjdz-uhfffaoysa, inchi=1/c3h5bro/c1-2-3 4 5/h2h2,1h3 | |
| propanoyl bromide |
Spécifications
| Propionyl bromide | |
| 1.5200g/mL | |
| 52°C | |
| 94% min. (GC) | |
| C3H5BrO | |
| CH3CH2COBr | |
| MFCD00000116 | |
| propionyl bromide, propanoyl-bromide, propionylbromide, propanyl bromide, acmc-209mg3, ksc270q9p, ribfxmjcuyxjdz-uhfffaoysa, inchi=1/c3h5bro/c1-2-3 4 5/h2h2,1h3 | |
| RIBFXMJCUYXJDZ-UHFFFAOYSA-N | |
| propanoyl bromide | |
| 69018 | |
| 95% |
| 598-22-1 | |
| 104°C | |
| Authentic | |
| Glass bottle | |
| 1.4525 to 1.4575 (20°C, 589nm) | |
| 25g | |
| 1.52 | |
| Solubility in water: reacts violently | |
| CCC(=O)Br | |
| 136.98 | |
| 136.98 |
Sécurité et manipulation
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 209-924-
Promotions