missing translation for 'onlineSavingsMsg'
Learn More

Propionamide, 97%

Catalog No. AC162670250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162670250 25 g Plastic bottle
AC162671000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC162670250 Supplier Thermo Scientific Chemicals Supplier No. 162670250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-05-0 | C3H7NO | 73.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-05-0
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.09
MDL Number MFCD00008039
InChI Key QLNJFJADRCOGBJ-UHFFFAOYSA-N
Synonym propionamide, propylamide, propionic amide, propionimidic acid, propanimidic acid, propionic acid amide, amid kyseliny propionove, propanimidic acid van, propionimidic acid van, unii-qk07g0hp47
PubChem CID 6578
ChEBI CHEBI:45422
IUPAC Name propanamide
SMILES CCC(=O)N

Specifications

Melting Point 79.0°C to 83.0°C
Color White to Yellow
Boiling Point 213.0°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula C2H5CONH2
Quantity 25 g
Beilstein 02, 243
Merck Index 15, 7938
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether, chloroform
Formula Weight 73.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Propionamide, 97%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-172-1
RTECSNumber UE2975000
TSCA TSCA

RUO – Research Use Only