missing translation for 'onlineSavingsMsg'
Learn More

Propiolamide, 96%

Catalog No. AAH52319MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52319MD 250 mg
AAH5231903 1 g
AAH5231906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52319MD Supplier Thermo Scientific Chemicals Supplier No. H52319MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7341-96-0 | C3H3NO | 69.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7341-96-0
Molecular Formula C3H3NO
Molecular Weight (g/mol) 69.06
MDL Number MFCD04035573
InChI Key HCJTYESURSHXNB-UHFFFAOYSA-N
Synonym propiolamide, 2-propynamide, propynoic acid amide, propynamide, propiolic acid amide, propargylamide, hc.equiv.cconh2, propynoicacidamide, 2-propynamide 9ci
PubChem CID 101445
ChEBI CHEBI:51764
IUPAC Name prop-2-ynamide
SMILES NC(=O)C#C

Specifications

Linear Formula HC≡CCONH2
Quantity 250 mg
Formula Weight 69.06
Percent Purity 96%
Chemical Name or Material Propiolamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only