missing translation for 'onlineSavingsMsg'
Learn More

Propargylaldehyde Diethyl Acetal 97.0+%, TCI America™

Catalog No. P11735ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11735ML 5 mL
P117325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11735ML Supplier TCI America Supplier No. P11735ML

Chemical Identifiers

CAS 10160-87-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00009237
InChI Key RGUXEWWHSQGVRZ-UHFFFAOYSA-N
Synonym 3,3-diethoxy-1-propyne, propargylaldehyde diethyl acetal, 3,3-diethoxypropyne, propiolaldehyde diethyl acetal, 1-propyne, 3,3-diethoxy, 3,3-diethoxy-propyne, propynal diethyl acetal, 1,1-diethoxyprop-2-yne, 1-propyne,3,3-diethoxy, propargyl aldehyde diethylacetal
PubChem CID 66285
IUPAC Name 3,3-diethoxyprop-1-yne
SMILES CCOC(OCC)C#C

Specifications

Color Yellow
Boiling Point 139°C
Quantity 5 mL
UN Number 3271
Formula Weight 128.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Propargylaldehyde Diethyl Acetal
TSCA No
Recommended Storage Refrigerator