missing translation for 'onlineSavingsMsg'
Learn More

Propargyl ether, 98%

Numéro de catalogue. p-7035746
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAH5347306 5 g
AAH5347314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAH5347306 Fournisseur Thermo Scientific Chemicals Code fournisseur H5347306
Il en reste null

CAS: 6921-27-3 | C6H6O | 94.113 g/mol

Propargyl ether is a useful ether for proteomics research. It was used for the preparation of polyferrocenes, via polyaddition with 1,1?-bis(azidoethyl)ferrocene. It may be used for the preaparation of cyclic polystyrene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl ether is a useful ether for proteomics research. It was used for the preparation of polyferrocenes, via polyaddition with 1,1′-bis(azidoethyl)ferrocene. It may be used for the preaparation of cyclic polystyrene.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Identifiants chimiques

CAS 6921-27-3
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.113
MDL Number MFCD00048108
InChI Key HRDCVMSNCBAMAM-UHFFFAOYSA-N
Synonym propargyl ether, dipropargyl ether, 2-propynyl ether, 1-propyne, 3,3'-oxybis, bispropargyl ether, 3-prop-2-yn-1-yloxy prop-1-yne, propargyl-ether, dipropargyl oxide, di 2-propynyl ether, bis 2-propynyl ether
PubChem CID 23349
IUPAC Name 3-prop-2-ynoxyprop-1-yne
SMILES C#CCOCC#C

Spécifications

Density 0.914
Boiling Point 55°C to 57°C (65 mmHg)
Flash Point 24°C (75°F)
Refractive Index 1.443
Linear Formula (HC≡CCH2)2O
Quantity 5 g
UN Number UN3271
Formula Weight 94.11
Percent Purity 98%
Chemical Name or Material Propargyl ether
Hazard Category H226-H302
Hazard Statement GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
RTECSNumber UK5950000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only