Learn More
Promega Wizard™ Plus SV Minipreps DNA Purification System
A silica membrane-based system for simple, rapid isolation of plasmid DNA from 1-10ml E. coli cultures. The miniprep procedure can be completed in 45 minutes or less.
Supplier:  Promega A1470
Encompass
 Afficher plus de versions de ce produit
Numéro de catalogue. PRA1470
Il en reste null
Comprend:
  • DNA purification kit: Cell resuspension solution, cell lysis solution, neutralization solution, column wash solution, alkaline protease solution, nuclease-free water, miniprep spin columns, collection tubes with or without vacuum adapters
  • DNA purification start-up kit (110V): Wizard™ Plus SV Minipreps DNA Purification System, Welch™ vacuum pump, Vac-Man™ laboratory vacuum manifold, vacuum adapters
Description
Description
  • 1-20μg High-Quality Plasmid DNA from 1-10mL Cultures
  • Process up to 20 minipreps in 45 minutes
  • Choice of spin or vacuum purification formats
  • No phenol extractions or precipitations required
Spécifications
Identifiants chimiques
15°C to 30°C
Spin or Vacuum
Produces high yield plasmid DNA of consistent quality from bacterial cultures
1 to 20μg
Spécifications
15°C to 30°C
Produces high yield plasmid DNA of consistent quality from bacterial cultures
250 Preps
1 to 20μg
Spin or Vacuum
250 Wizard SV minicolumns, 250 Collection tubes, 100mL Column wash solution, 3mL Alkaline Protease solution, 100mL Neutralization solution, 75mL Cell resuspension solution, 75mL Cell Lysis solution, 25mL Nuclease-free water, 20 Vacuum adapters
DNA Purification, Vac Adapters
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
U.S. Patent 5,981,235, Australian Patent 729932.