Please login to your online account to display your discounted pricing

Promega™ Ultra Pure Deoxynucleotide Triphosphates (dNTPs)


High-quality dNTPs for cDNA synthesis, sequencing, and labeling

Manufacturer: promega™  U1205

 View more versions of this product

Catalog No. PR-U1205

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) cDNA synthesis, sequencing and labeling, can be used in place of dTTP in PCR and RT-PCR protocols
Includes Individual tubes of each dNTP
pH 7.5
Product Type dATP
Concentration 100mM
Purity >99%
Quantity 25μm
Storage Requirements -30° to -10°C
Product Type dATP