Learn More
Promega SV 96 Total RNA Isolation System
The SV 96 Total RNA Isolation System provides a high-throughput technique to prepare intact RNA from tissue and cultured cells.
Supplier:  Promega Z3505
Encompass
 View more versions of this product
Catalog No. PRZ3505
Only null left
Includes: Binding plate; DNA elution plate; DNase I (lyophilized); β-mercaptoethanol; SV DNase stop solution; MnCl2 (0.09M); Nuclease-free water; SV RNA wash solution; SV RNA lysis buffer; Yellow core buffer; Plate sealers
Requires : Use of the Vac-Man™ 96 Vacuum Manifold (Mfr. No. PR-A2291) or similar 96-well manifold for manual purification.
Description
Description
  • Purifies Total RNA From 96 Samples in Under an Hour
  • Vacuum-based method. No centrifugation required
  • Plate design prevents cross-contamination during sample processing
  • Designed for both manual and automated RNA purification
Specifications
Chemical Identifiers
15°C to 30°C
Preparation of intact, purified total RNA and RT-PCR
Specifications
15°C to 30°C
Preparation of intact, purified total RNA and RT-PCR
Binding plate 5 x 1 plate, elution plate 5 x 1 plate, plate sealers 2 x 3, Nuclease-free Water 5 x 13mL, RNA Lysis Buffer (RLA) 2 x 50mL, RNA wash solution (RWA) 5 x 58.8mL, DNase stop solution (DSA) 5 x 13.25mL, yellow core Buffer 5 x 2.5mL, MnCl2, 0.09M 5 x 750μL, DNase I (lyophilized) 5 x 1 vial, Beta-Mercaptoethanol 2 x 2mL
5 x 96 Isolations
Product Suggestions
Product Suggestions