missing translation for 'onlineSavingsMsg'
Learn More

Promega Streptavidin MagneSphere™ Paramagnetic Particles

Streptavidin MagneSphere Paramagnetic Particles (PMPs) are used for magnetic separation and purification of a variety of biotinylated nucleic acid or protein molecules.

$372.88 - $953.95

Specifications

Concentration 1mg/mL
Content And Storage 2°C to 10°C
For Use With (Application) Magnetic separation and purification of a wide variety of Biotinylated Nucleic Acid or Protein molecules
Product Type Paramagnetic Particles
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRZ5481
Encompass
View Documents Promotion Details
Promega
Z5481
9 mL
Each for $372.88
Only null left
 
PRZ5482
Encompass
View Documents Promotion Details
Promega
Z5482
25 mL
Each for $953.95
Only null left
 
Description

Description

  • Separation and Purification of Biotinylated Nucleic Acids or Proteins
  • Strong, specific binding to biotinylated molecules
  • Enable binding, washing and magnetic separation from undesired materials
  • Used for purification of DNA, mRNA and proteins.
  • Supplied at a concentration of 1mg/ml
Specifications

Specifications

1mg/mL
2°C to 10°C
Magnetic separation and purification of a wide variety of Biotinylated Nucleic Acid or Protein molecules
Paramagnetic Particles
Videos
SDS
Documents

Documents