Learn More
Promega Streptavidin MagneSphere™ Paramagnetic Particles
Streptavidin MagneSphere Paramagnetic Particles (PMPs) are used for magnetic separation and purification of a variety of biotinylated nucleic acid or protein molecules.
Supplier:  Promega Z5482
Encompass
 Afficher plus de versions de ce produit
Numéro de catalogue. PRZ5482
Il en reste null
Description
Description
  • Separation and Purification of Biotinylated Nucleic Acids or Proteins
  • Strong, specific binding to biotinylated molecules
  • Enable binding, washing and magnetic separation from undesired materials
  • Used for purification of DNA, mRNA and proteins.
  • Supplied at a concentration of 1mg/ml
Spécifications
Identifiants chimiques
1mg/mL
2°C to 10°C
Magnetic separation and purification of a wide variety of Biotinylated Nucleic Acid or Protein molecules
Paramagnetic Particles
Spécifications
1mg/mL
Magnetic separation and purification of a wide variety of Biotinylated Nucleic Acid or Protein molecules
25 mL
2°C to 10°C
Paramagnetic Particles
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS